Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00005887.mm.1 |
NM_010833.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005887.mm.1 |
NM_010833.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005887.mm.1 |
TC0X00000916.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00005888.mm.1 |
NM_010833.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005888.mm.1 |
NM_010833.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005888.mm.1 |
TC0X00000916.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00005889.mm.1 |
NM_010833.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005889.mm.1 |
NM_010833.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005889.mm.1 |
TC0X00000916.mm.1 |
MTA TC |