Assay Details
| Gene Symbol: Sept2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010891.2 | NP_035021.1 | 2 - 3 | 107 | 86 | |
| GenBank mRNA | AK028072.1 | - | 2 - 3 | 89 | 86 | |
| AK146616.1 | - | 2 - 3 | 104 | 86 | ||
| AK151591.1 | - | 2 - 3 | 91 | 86 | ||
| AK171331.1 | - | 2 - 3 | 86 | 86 | ||
| BC138636.1 | - | 1 - 2 | 44 | 8861656 | 86 | |
| BC138637.1 | - | 1 - 2 | 44 | 8861657 | 86 | |
| CJ075082.1 | - | 2 - 3 | 130 | 86 | ||
| D49382.2 | - | 2 - 3 | 62 | 86 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK028072.1,AK146616.1,AK151591.1,AK171331.1,BC138636.1,BC138637.1,D49382.2,NM_010891.2 transcripts.
Set Membership: |
|

Sept2
Genomic Map