Assay Details
| Gene Symbol: Nfia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122952.1 | NP_001116424.1 | 5 - 6 | 927 | 63 | |
| NM_001122953.1 | NP_001116425.1 | 5 - 6 | 1166 | 63 | ||
| NM_010905.3 | NP_035035.1 | 5 - 6 | 1166 | 63 | ||
| GenBank mRNA | AF326553.1 | - | 5 - 6 | 865 | 63 | |
| AF326554.1 | - | 5 - 6 | 865 | 63 | ||
| AK050320.1 | - | 4 - 5 | 483 | 63 | ||
| AK052204.1 | - | 5 - 6 | 988 | 63 | ||
| AK054310.1 | - | 5 - 6 | 1030 | 63 | ||
| AK137731.1 | - | 5 - 6 | 1029 | 63 | ||
| AK139357.1 | - | 5 - 6 | 928 | 63 | ||
| AK173173.1 | - | 4 - 5 | 802 | 63 | ||
| BC075702.1 | - | 5 - 6 | 1147 | 30087137 | 63 | |
| BC122878.1 | - | 5 - 6 | 888 | 40107695 | 63 | |
| BC122880.1 | - | 5 - 6 | 886 | 63 | ||
| D90172.1 | - | 5 - 6 | 1096 | 63 | ||
| D90173.1 | - | 5 - 6 | 1004 | 63 | ||
| D90174.1 | - | 5 - 6 | 1037 | 63 | ||
| D90175.1 | - | 881 | 63 | |||
| D90176.1 | - | 5 - 6 | 919 | 63 | ||
| U57633.1 | - | 5 - 6 | 821 | 63 | ||
| Y07690.1 | - | 5 - 6 | 897 | 63 | ||
| Y07691.1 | - | 5 - 6 | 895 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfia
Genomic Map