Assay Details
| Gene Symbol: Nfia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122952.1 | NP_001116424.1 | 7 - 8 | 1177 | 56 | |
| NM_010905.3 | NP_035035.1 | 7 - 8 | 1416 | 56 | ||
| GenBank mRNA | AF326553.1 | - | 7 - 8 | 1115 | 56 | |
| AF326554.1 | - | 7 - 8 | 1115 | 56 | ||
| AK052204.1 | - | 7 - 8 | 1238 | 56 | ||
| AK137731.1 | - | 7 - 8 | 1279 | 56 | ||
| AK139357.1 | - | 7 - 8 | 1178 | 56 | ||
| AK173173.1 | - | 6 - 7 | 1052 | 56 | ||
| BC075702.1 | - | 7 - 8 | 1397 | 30087137 | 56 | |
| D90172.1 | - | 7 - 8 | 1346 | 56 | ||
| D90173.1 | - | 7 - 8 | 1254 | 56 | ||
| D90174.1 | - | 7 - 8 | 1287 | 56 | ||
| D90175.1 | - | 1131 | 56 | |||
| U57633.1 | - | 7 - 8 | 1071 | 56 | ||
| Y07690.1 | - | 7 - 8 | 1147 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfia
Genomic Map