Assay Details
| Gene Symbol: Nrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164226.1 | NP_001157698.1 | 4 - 5 | 632 | 69 | |
| NM_001164227.1 | NP_001157699.1 | 4 - 5 | 632 | 69 | ||
| NM_001164228.1 | NP_001157700.1 | 4 - 5 | 629 | 69 | ||
| NM_001164229.1 | NP_001157701.1 | 5 - 6 | 706 | 69 | ||
| NM_001164230.1 | NP_001157702.1 | 4 - 5 | 629 | 69 | ||
| NM_010938.4 | NP_035068.3 | 4 - 5 | 629 | 69 | ||
| GenBank mRNA | AF098077.1 | - | 4 - 5 | 564 | 69 | |
| AK014494.1 | - | 4 - 5 | 593 | 69 | ||
| AK029034.1 | - | 4 - 5 | 610 | 69 | ||
| AK037697.1 | - | 4 - 5 | 590 | 69 | ||
| AK050861.1 | - | 4 - 5 | 546 | 69 | ||
| AK082580.1 | - | 4 - 5 | 630 | 69 | ||
| AK134483.1 | - | 4 - 5 | 546 | 69 | ||
| AK135673.1 | - | 5 - 6 | 709 | 69 | ||
| BC005410.1 | - | 3 - 4 | 450 | 3496661 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nrf1
Genomic Map