Assay Details
Gene Symbol: Nrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164226.1 | NP_001157698.1 | 11 - 12 | 1640 | 67 | |
NM_010938.4 | NP_035068.3 | 11 - 12 | 1637 | 67 | ||
GenBank mRNA | AF098077.1 | - | 11 - 12 | 1572 | 67 | |
AK014494.1 | - | 11 - 12 | 1601 | 67 | ||
AK029034.1 | - | 11 - 12 | 1618 | 67 | ||
AK031103.1 | - | 6 - 7 | 841 | 67 | ||
AK082580.1 | - | 11 - 12 | 1638 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
