Assay Details
| Gene Symbol: Pltp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011125.2 | NP_035255.1 | 4 - 5 | 464 | 59 | |
| GenBank mRNA | AK148330.1 | - | 4 - 5 | 469 | 59 | |
| AK149730.1 | - | 4 - 5 | 395 | 59 | ||
| AK190857.1 | - | 1 - 2 | 128 | 59 | ||
| BC003782.1 | - | 4 - 5 | 398 | 3491360 | 59 | |
| CT010317.1 | - | 3 - 4 | 328 | 59 | ||
| U37226.1 | - | 4 - 5 | 394 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pltp
Genomic Map