Assay Details
| Gene Symbol: Sept4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284392.1 | NP_001271321.1 | 8 - 9 | 926 | 117 | |
| NM_001284394.1 | NP_001271323.1 | 9 - 10 | 1223 | 117 | ||
| NM_001284398.1 | NP_001271327.1 | 8 - 9 | 910 | 117 | ||
| NM_011129.2 | NP_035259.1 | 9 - 10 | 1223 | 117 | ||
| GenBank mRNA | AB078010.1 | - | 8 - 9 | 821 | 117 | |
| AK133933.1 | - | 8 - 9 | 878 | 117 | ||
| AK135143.1 | - | 9 - 10 | 1224 | 117 | ||
| BC055101.1 | - | 9 - 10 | 1223 | 6591016 | 117 | |
| X61452.1 | - | 9 - 10 | 1153 | 117 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sept4
Genomic Map