Assay Details
| Gene Symbol: Pold1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011131.3 | NP_035261.3 | 5 - 6 | 647 | 59 | |
| GenBank mRNA | AK088310.1 | - | 5 - 6 | 638 | 59 | |
| AK146837.1 | - | 5 - 6 | 643 | 59 | ||
| AK166592.1 | - | 5 - 6 | 841 | 59 | ||
| AK167569.1 | - | 5 - 6 | 630 | 59 | ||
| AK168967.1 | - | 5 - 6 | 610 | 59 | ||
| AK169040.1 | - | 5 - 6 | 643 | 59 | ||
| BC009128.1 | - | 5 - 6 | 648 | 3257174 | 59 | |
| BC052670.1 | - | 4 - 5 | 614 | 30058314 | 59 | |
| Z21848.1 | - | 5 - 6 | 616 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pold1
Genomic Map