Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100028341.mm.1 |
NM_011137.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100028341.mm.1 |
NM_011137.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100028341.mm.1 |
NM_198932.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100028341.mm.1 |
NM_198932.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100028341.mm.1 |
NM_198933.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100028341.mm.1 |
NM_198933.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100028341.mm.1 |
NM_198934.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100028341.mm.1 |
NM_198934.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100028341.mm.1 |
TC0100003457.mm.1 |
MTA TC |