Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500028314.mm.1 |
TC0500003184.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500028314.mm.1 |
NM_001109992.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028314.mm.1 |
NM_001109992.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028314.mm.1 |
NM_011202.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028314.mm.1 |
NM_011202.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028316.mm.1 |
TC0500003184.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500028316.mm.1 |
NM_001109992.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028316.mm.1 |
NM_001109992.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028316.mm.1 |
NM_011202.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028316.mm.1 |
NM_011202.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028315.mm.1 |
TC0500003184.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500028315.mm.1 |
NM_001109992.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028315.mm.1 |
NM_001109992.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028315.mm.1 |
NM_011202.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028315.mm.1 |
NM_011202.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028317.mm.1 |
TC0500003184.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500028317.mm.1 |
NM_001109992.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028317.mm.1 |
NM_001109992.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028317.mm.1 |
NM_011202.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028317.mm.1 |
NM_011202.3 |
Refseq |