Assay Details
| Gene Symbol: Ptpre | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011212.3 | NP_035342.3 | 9 - 10 | 946 | 90 | |
| GenBank mRNA | AK004602.1 | - | 9 - 10 | 946 | 90 | |
| AK132455.1 | - | 8 - 9 | 909 | 90 | ||
| AK154910.1 | - | 6 - 7 | 498 | 90 | ||
| AK160947.1 | - | 6 - 7 | 495 | 90 | ||
| BC053408.1 | - | 9 - 10 | 722 | 90 | ||
| BC098473.1 | - | 9 - 10 | 710 | 90 | ||
| D83484.1 | - | 9 - 10 | 1283 | 90 | ||
| U35368.1 | - | 9 - 10 | 946 | 90 | ||
| U36758.1 | - | 6 - 7 | 495 | 90 | ||
| U40280.1 | - | 8 - 9 | 686 | 90 | ||
| U62387.1 | - | 9 - 10 | 848 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpre
Genomic Map