Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000009968.mm.1 |
NM_001161837.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009968.mm.1 |
NM_001161837.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009968.mm.1 |
NM_001161838.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009968.mm.1 |
NM_001161838.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009968.mm.1 |
NM_001161839.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009968.mm.1 |
NM_001161839.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009968.mm.1 |
NM_001161840.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009968.mm.1 |
NM_001161840.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009968.mm.1 |
NM_011217.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009968.mm.1 |
NM_011217.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009968.mm.1 |
TC1000001334.mm.1 |
MTA TC |