Assay Details
| Gene Symbol: Uri1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011274.5 | NP_035404.4 | 7 - 8 | 1018 | 69 | |
| GenBank mRNA | AF091096.1 | - | 5 - 6 | 540 | 69 | |
| AK052034.1 | - | 2 - 3 | 199 | 69 | ||
| AK133383.1 | - | 7 - 8 | 693 | 69 | ||
| AK155994.1 | - | 7 - 8 | 992 | 69 | ||
| AK162285.1 | - | 2 - 3 | 192 | 69 | ||
| AK166566.1 | - | 7 - 8 | 1009 | 69 | ||
| AK170929.1 | - | 7 - 8 | 1013 | 69 | ||
| BC023029.1 | - | 7 - 8 | 678 | 5374731 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Uri1
Genomic Map