Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700016153.mm.1 |
NM_001098226.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016153.mm.1 |
NM_001098226.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016153.mm.1 |
NM_001204156.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016153.mm.1 |
NM_001204156.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016153.mm.1 |
NM_011566.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016153.mm.1 |
NM_011566.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016153.mm.1 |
TC1700001755.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700016155.mm.1 |
NM_001098226.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016155.mm.1 |
NM_001098226.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016155.mm.1 |
NM_001204156.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016155.mm.1 |
NM_001204156.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016155.mm.1 |
NM_011566.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016155.mm.1 |
NM_011566.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016155.mm.1 |
TC1700001755.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700016146.mm.1 |
NM_001098226.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016146.mm.1 |
NM_001098226.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016146.mm.1 |
NM_001204156.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016146.mm.1 |
NM_001204156.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016146.mm.1 |
NM_011566.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016146.mm.1 |
NM_011566.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016146.mm.1 |
TC1700001755.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700016147.mm.1 |
NM_001098226.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016147.mm.1 |
NM_001098226.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016147.mm.1 |
NM_001204156.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016147.mm.1 |
NM_001204156.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016147.mm.1 |
NM_011566.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016147.mm.1 |
NM_011566.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016147.mm.1 |
TC1700001755.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700016156.mm.1 |
NM_001204156.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016156.mm.1 |
NM_001204156.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016156.mm.1 |
TC1700001755.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700016160.mm.1 |
NM_011566.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016160.mm.1 |
NM_011566.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016160.mm.1 |
TC1700001755.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700016154.mm.1 |
NM_001098226.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016154.mm.1 |
NM_001098226.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016154.mm.1 |
NM_001204156.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016154.mm.1 |
NM_001204156.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016154.mm.1 |
NM_011566.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016154.mm.1 |
NM_011566.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016154.mm.1 |
TC1700001755.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700016159.mm.1 |
NM_011566.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016159.mm.1 |
NM_011566.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016159.mm.1 |
TC1700001755.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700016157.mm.1 |
NM_001098226.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016157.mm.1 |
NM_001098226.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016157.mm.1 |
TC1700001755.mm.1 |
MTA TC |