Assay Details
| Gene Symbol: Vcam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011693.3 | NP_035823.3 | 3 - 4 | 979 | 79 | |
| GenBank mRNA | AK016465.1 | - | 3 - 4 | 748 | 79 | |
| AK030195.1 | - | 3 - 4 | 749 | 79 | ||
| AK143401.1 | - | 3 - 4 | 749 | 79 | ||
| AK162954.1 | - | 3 - 4 | 749 | 79 | ||
| BC029823.1 | - | 3 - 4 | 979 | 4196681 | 79 | |
| CT010373.1 | - | 3 - 4 | 658 | 79 | ||
| L08431.1 | - | 3 - 4 | 718 | 79 | ||
| M84487.1 | - | 3 - 4 | 715 | 79 | ||
| U12884.1 | - | 3 - 4 | 701 | 79 | ||
| X67783.1 | - | 3 - 4 | 673 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vcam1
Genomic Map