| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700000778.mm.1 |
NM_011866.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000778.mm.1 |
NM_011866.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000778.mm.1 |
TC1700000092.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700000779.mm.1 |
NM_011866.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000779.mm.1 |
NM_011866.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000779.mm.1 |
TC1700000092.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700000786.mm.1 |
NM_011866.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000786.mm.1 |
NM_011866.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000786.mm.1 |
TC1700000092.mm.1 |
MTA TC |