Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400011186.mm.1 |
NM_011970.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011186.mm.1 |
NM_011970.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011186.mm.1 |
TC0400001469.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400011187.mm.1 |
NM_011970.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011187.mm.1 |
NM_011970.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011187.mm.1 |
TC0400001469.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400011190.mm.1 |
NM_011970.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011190.mm.1 |
NM_011970.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011190.mm.1 |
TC0400001469.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400011189.mm.1 |
NM_011970.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011189.mm.1 |
NM_011970.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011189.mm.1 |
TC0400001469.mm.1 |
MTA TC |