Assay Details
| Gene Symbol: Nit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242580.1 | NP_001229509.1 | 3 - 4 | 447 | 82 | |
| NM_012049.2 | NP_036179.1 | 3 - 4 | 415 | 82 | ||
| GenBank mRNA | AF069988.1 | - | 3 - 4 | 396 | 82 | |
| AK004988.2 | - | 408 | 82 | |||
| AK200650.1 | - | 2 - 3 | 295 | 82 | ||
| AK205756.1 | - | 2 - 3 | 295 | 82 | ||
| BC021634.1 | - | 3 - 4 | 397 | 4008543 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nit1
Genomic Map