Assay Details
| Gene Symbol: Supt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013676.1 | NP_038704.1 | 9 - 10 | 585 | 88 | |
| GenBank mRNA | AK047062.1 | - | 1 - 2 | 69 | 88 | |
| AK145117.1 | - | 9 - 10 | 625 | 88 | ||
| AK145465.1 | - | 9 - 10 | 577 | 88 | ||
| AK146055.1 | - | 9 - 10 | 613 | 88 | ||
| AK146453.1 | - | 9 - 10 | 613 | 88 | ||
| AK146583.1 | - | 8 - 9 | 832 | 88 | ||
| AK146650.1 | - | 9 - 10 | 626 | 88 | ||
| AK159786.1 | - | 10 - 11 | 710 | 88 | ||
| BC007132.1 | - | 9 - 10 | 585 | 3496691 | 88 | |
| BC057449.1 | - | 9 - 10 | 600 | 88 | ||
| BC058598.1 | - | 8 - 9 | 560 | 6306109 | 88 | |
| BC059849.1 | - | 1 - 2 | 168 | 5696323 | 88 | |
| U88539.1 | - | 9 - 10 | 612 | 88 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC059849.1 transcripts.
Set Membership: |
|

Supt5
Genomic Map