Assay Details
| Gene Symbol: Skp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285980.1 | NP_001272909.1 | 7 - 8 | 1087 | 80 | |
| NM_013787.3 | NP_038815.1 | 7 - 8 | 1157 | 80 | ||
| GenBank mRNA | AF083215.1 | - | 7 - 8 | 1020 | 80 | |
| AK006221.1 | - | 7 - 8 | 1087 | 80 | ||
| AK032979.1 | - | 7 - 8 | 1068 | 80 | ||
| AK044212.1 | - | 7 - 8 | 1058 | 80 | ||
| AK166034.1 | - | 7 - 8 | 1058 | 80 | ||
| AK167677.1 | - | 7 - 8 | 1055 | 80 | ||
| BC092236.1 | - | 7 - 8 | 1031 | 30610417 | 80 | |
| BC098371.1 | - | 7 - 8 | 1140 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Skp2
Genomic Map