Assay Details
Gene Symbol: Preb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001294302.1 | NP_001281231.1 | 2 - 3 | 544 | 103 | |
NM_016703.3 | NP_057912.2 | 2 - 3 | 544 | 103 | ||
GenBank mRNA | AF150808.1 | - | 2 - 3 | 447 | 103 | |
AF193017.1 | - | 2 - 3 | 459 | 103 | ||
AK013330.1 | - | 2 - 3 | 498 | 103 | ||
AK029838.1 | - | 2 - 3 | 479 | 103 | ||
AK036866.1 | - | 2 - 3 | 482 | 103 | ||
AK151288.1 | - | 2 - 3 | 526 | 103 | ||
BC017527.1 | - | 2 - 3 | 447 | 4921119 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
