Assay Details
Gene Symbol: Preb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001294302.1 | NP_001281231.1 | 3 - 4 | 770 | 64 | |
NM_016703.3 | NP_057912.2 | 3 - 4 | 770 | 64 | ||
GenBank mRNA | AF150808.1 | - | 3 - 4 | 673 | 64 | |
AF193017.1 | - | 3 - 4 | 682 | 64 | ||
AK013330.1 | - | 3 - 4 | 724 | 64 | ||
AK029838.1 | - | 3 - 4 | 705 | 64 | ||
AK151288.1 | - | 3 - 4 | 752 | 64 | ||
AK213634.1 | - | 1 - 2 | 85 | 64 | ||
BC017527.1 | - | 3 - 4 | 673 | 4921119 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
