Assay Details
| Gene Symbol: Tnk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110147.1 | NP_001103617.1 | 6 - 7 | 1370 | 60 | |
| NM_016788.3 | NP_058068.2 | 6 - 7 | 1370 | 60 | ||
| GenBank mRNA | AF037260.1 | - | 6 - 7 | 1153 | 60 | |
| AK087965.1 | - | 3 - 4 | 527 | 60 | ||
| BC052421.1 | - | 6 - 7 | 1370 | 6400209 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tnk2
Genomic Map