Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00011557.mm.1 |
NM_016913.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011557.mm.1 |
NM_023638.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011557.mm.1 |
NM_145907.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011557.mm.1 |
NM_145908.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011557.mm.1 |
TC0X00001800.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00011556.mm.1 |
NM_016913.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011556.mm.1 |
NM_023638.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011556.mm.1 |
NM_145907.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011556.mm.1 |
NM_145908.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011556.mm.1 |
TC0X00001800.mm.1 |
MTA TC |