Assay Details
Gene Symbol: Stag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016964.2 | NP_058660.2 | 6 - 7 | 713 | 71 | |
GenBank mRNA | AJ005678.1 | - | 6 - 7 | 671 | 71 | |
AK145898.1 | - | 6 - 7 | 699 | 71 | ||
AK158240.1 | - | 6 - 7 | 609 | 71 | ||
BC139010.1 | - | 6 - 7 | 643 | 8862032 | 71 | |
BC139011.1 | - | 6 - 7 | 643 | 8862033 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
