Assay Details
| Gene Symbol: Rfx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017395.2 | NP_059091.2 | 1 - 2 | 136 | 100 | |
| GenBank mRNA | AB221679.1 | - | 1 - 2 | 117 | 100 | |
| AF209854.1 | - | 1 - 2 | 117 | 100 | ||
| AK088686.1 | - | 2 - 3 | 188 | 100 | ||
| BB854843.1 | - | 2 - 3 | 229 | 100 | ||
| BC057336.1 | - | 2 - 3 | 162 | 100 | ||
| BC075717.1 | - | 3 - 4 | 245 | 100 | ||
| BC118948.2 | - | 1 - 2 | 117 | 40092396 | 100 | |
| BC118949.2 | - | 1 - 2 | 117 | 40092397 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rfx5
Genomic Map