Assay Details
| Gene Symbol: Rfx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017395.2 | NP_059091.2 | 5 - 6 | 494 | 85 | |
| GenBank mRNA | AB221679.1 | - | 5 - 6 | 475 | 85 | |
| AF209854.1 | - | 5 - 6 | 475 | 85 | ||
| BB666132.1 | - | 6 - 7 | 520 | 85 | ||
| BC051965.1 | - | 6 - 7 | 531 | 5702640 | 85 | |
| BC057336.1 | - | 6 - 7 | 520 | 85 | ||
| BC075717.1 | - | 7 - 8 | 603 | 85 | ||
| BC118948.2 | - | 5 - 6 | 475 | 40092396 | 85 | |
| BC118949.2 | - | 5 - 6 | 475 | 40092397 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rfx5
Genomic Map