Assay Details
| Gene Symbol: Polm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017401.2 | NP_059097.2 | 10 - 11 | 1668 | 114 | |
| GenBank mRNA | AF176098.1 | - | 10 - 11 | 1441 | 114 | |
| AJ251804.2 | - | 10 - 11 | 1412 | 114 | ||
| BC056213.1 | - | 10 - 11 | 1460 | 30024844 | 114 | |
| BC056996.1 | - | 7 - 8 | 3315 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
DNA repair
DNA recombination
cellular response to DNA damage stimulus
somatic hypermutation of immunoglobulin genes
B cell differentiation
DNA binding
DNA-directed DNA polymerase activity
transferase activity
nucleotidyltransferase activity
DNA polymerase activity
metal ion binding
DNA repair
DNA recombination
cellular response to DNA damage stimulus
somatic hypermutation of immunoglobulin genes
B cell differentiation
DNA binding
DNA-directed DNA polymerase activity
transferase activity
nucleotidyltransferase activity
DNA polymerase activity
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27565172 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27565178 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27565173 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27565177 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27565176 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27565179 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27565175 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042590 | BC056213.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042590 | BC056996.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042590 | AJ251804.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042590 | NM_017401.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2122255 | AJ251804.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 611049 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 588730 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 821128 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 67686 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 552775 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 411153 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 486874 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 111980 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 23056 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105045 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042590 | BC056213.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042590 | BC056996.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042590 | AJ251804.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042590 | NM_017401.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2122255 | AJ251804.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P397493 | BC056213.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P397493 | BC056996.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P397493 | AJ251804.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P397493 | NM_017401.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565172 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565200 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565191 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565179 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565161 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565180 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565201 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105045 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07244205 | - | - |
| Whole Mouse Genome G4122A | A_51_P397493 | BC056213.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P397493 | BC056996.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P397493 | AJ251804.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P397493 | NM_017401.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113618 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105045 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09189590 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09050825 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09050819 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09050816 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09050823 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27565176 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27565172 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27565179 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27565178 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27565175 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27565173 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27565177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09078984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09329931 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09346866 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09329925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09050828 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09050825 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565136 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09447291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09329930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09467996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09189590 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565169 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565140 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565133 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565208 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09050816 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565210 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565139 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565137 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565172 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09447294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565161 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09203073 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565203 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10384025 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384028 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384022 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384029 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384031 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384027 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384023 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384026 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384030 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384024 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384021 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417249_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10384020 | - | - |
| Mouse Genome 430 2.0 Array | 1417249_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417249_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 54125_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 54125_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100020909.mm.1 | NM_017401.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100020909.mm.1 | NM_017401.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100020909.mm.1 | TC1100002154.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417249_PM_at | - | - |
| Mouse Expression Set 430 | 1417249_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417249_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417249_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417249_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417249_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 659293 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2630440 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002630440 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2621761 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2621761 | - | - |
| mouse-6 v1.1 (Illumina) | scl40553.9.1_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2630440 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40553.9.1_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2630440 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2621761 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2621762 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2621762 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2621761 | - | - |

Polm
Genomic Map