Assay Details
| Gene Symbol: Mre11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018736.2 | NP_061206.1 | 6 - 7 | 676 | 67 | |
| GenBank mRNA | AK041248.1 | - | 6 - 7 | 675 | 67 | |
| AK141214.1 | - | 6 - 7 | 688 | 67 | ||
| AK146234.1 | - | 6 - 7 | 709 | 67 | ||
| AK146370.1 | - | 6 - 7 | 709 | 67 | ||
| AK168180.1 | - | 6 - 7 | 709 | 67 | ||
| AK168208.1 | - | 6 - 7 | 710 | 67 | ||
| BC065144.1 | - | 6 - 7 | 676 | 30627207 | 67 | |
| U58987.1 | - | 6 - 7 | 711 | 67 | ||
| U60318.1 | - | 6 - 7 | 711 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mre11a
Genomic Map