Assay Details
| Gene Symbol: Mre11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018736.2 | NP_061206.1 | 12 - 13 | 1453 | 90 | |
| GenBank mRNA | AK041248.1 | - | 12 - 13 | 1452 | 90 | |
| AK141214.1 | - | 12 - 13 | 1465 | 90 | ||
| AK146234.1 | - | 11 - 12 | 1405 | 90 | ||
| AK146370.1 | - | 11 - 12 | 1405 | 90 | ||
| AK168180.1 | - | 11 - 12 | 1405 | 90 | ||
| AK168208.1 | - | 12 - 13 | 1488 | 90 | ||
| BC065144.1 | - | 12 - 13 | 1453 | 30627207 | 90 | |
| U58987.1 | - | 12 - 13 | 1488 | 90 | ||
| U60318.1 | - | 11 - 12 | 1407 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mre11a
Genomic Map