Assay Details
| Gene Symbol: Mre11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018736.2 | NP_061206.1 | 17 - 18 | 2058 | 95 | |
| GenBank mRNA | AK141214.1 | - | 17 - 18 | 2070 | 95 | |
| AK161013.1 | - | 5 - 6 | 524 | 95 | ||
| AK168208.1 | - | 17 - 18 | 2093 | 95 | ||
| BC065144.1 | - | 17 - 18 | 2058 | 30627207 | 95 | |
| U58987.1 | - | 17 - 18 | 2093 | 95 | ||
| U60318.1 | - | 16 - 17 | 2012 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mre11a
Genomic Map