Assay Details
| Gene Symbol: Ppt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK195061.1 | - | 2 - 3 | 163 | 77 | |
| BC013462.1 | - | 5 - 6 | 760 | 4238225 | 77 | |
| BC052330.1 | - | 6 - 7 | 734 | 77 | ||
| CX201936.1 | - | 3 - 4 | 214 | 77 | ||
| CX217389.1 | - | 6 - 7 | 698 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppt2
Genomic Map