Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200001736.mm.1 |
NM_019501.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200001736.mm.1 |
NM_019501.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200001736.mm.1 |
TC0200000354.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200001737.mm.1 |
NM_019501.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200001737.mm.1 |
NM_019501.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200001737.mm.1 |
TC0200000354.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200001735.mm.1 |
NM_019501.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200001735.mm.1 |
NM_019501.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200001735.mm.1 |
TC0200000354.mm.1 |
MTA TC |