Assay Details
Gene Symbol: Pdss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019501.3 | NP_062374.2 | 4 - 5 | 559 | 98 | |
GenBank mRNA | AB210841.1 | - | 4 - 5 | 479 | 98 | |
AF118855.1 | - | 2 - 3 | 254 | 98 | ||
AK012307.1 | - | 4 - 5 | 487 | 98 | ||
BC026820.1 | - | 4 - 5 | 461 | 4039464 | 98 | |
BC107273.1 | - | 4 - 5 | 374 | 98 | ||
BC107274.1 | - | 4 - 5 | 378 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
