Assay Details
| Gene Symbol: Rev1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019570.3 | NP_062516.2 | 4 - 5 | 565 | 71 | |
| GenBank mRNA | AB057418.1 | - | 3 - 4 | 354 | 71 | |
| AF179302.1 | - | 3 - 4 | 354 | 71 | ||
| AK048838.1 | - | 4 - 5 | 570 | 71 | ||
| BC058093.1 | - | 4 - 5 | 565 | 6827108 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rev1
Genomic Map