Assay Details
| Gene Symbol: Ptpn9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019651.2 | NP_062625.2 | 1 - 2 | 688 | 90 | |
| GenBank mRNA | AF013490.1 | - | 1 - 2 | 90 | ||
| AK089069.1 | - | 1 - 2 | 261 | 90 | ||
| BC053017.1 | - | 1 - 2 | 688 | 5708923 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn9
Genomic Map