Assay Details
| Gene Symbol: Rrad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019662.2 | NP_062636.2 | 2 - 3 | 634 | 71 | |
| GenBank mRNA | AF084466.1 | - | 1 - 2 | 377 | 71 | |
| AK077271.1 | - | 2 - 3 | 632 | 71 | ||
| AK087473.1 | - | 2 - 3 | 607 | 71 | ||
| AK159550.1 | - | 1 - 2 | 417 | 71 | ||
| BC057138.1 | - | 2 - 3 | 613 | 5149047 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26129565 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26129568 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26129564 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26129566 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240253 | BC057138.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240253 | AF084466.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240253 | AK077271.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240253 | AK087473.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240253 | AK159550.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240253 | BC057138.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240253 | AF084466.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240253 | AK077271.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240253 | AK087473.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240253 | AK159550.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 597058 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 889369 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 67753 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123807 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240253 | BC057138.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240253 | AF084466.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240253 | AK077271.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240253 | AK087473.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240253 | AK159550.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240253 | BC057138.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240253 | AF084466.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240253 | AK077271.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240253 | AK087473.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240253 | AK159550.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240253 | BC057138.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240253 | AF084466.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240253 | AK077271.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240253 | AK087473.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240253 | AK159550.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240253 | BC057138.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240253 | AF084466.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240253 | AK077271.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240253 | AK087473.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240253 | AK159550.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129595 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129577 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129592 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129599 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129562 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129590 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129582 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129600 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129581 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129584 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129561 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129588 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129559 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129570 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129565 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129558 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129568 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129597 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129575 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129587 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129578 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129593 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129563 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129598 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26129572 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P123807 | - | - |
| Whole Mouse Genome G4122A | A_51_P240253 | BC057138.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P240253 | AF084466.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P240253 | AK077271.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P240253 | AK087473.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P240253 | AK159550.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P240253 | BC057138.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P240253 | AF084466.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P240253 | AK077271.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P240253 | AK087473.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P240253 | AK159550.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131397 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105701 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P123807 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07088279 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07319290 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07088278 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07319275 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07088277 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06968672 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07088269 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07319288 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26129565 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26129566 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26129568 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26129564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26129609 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129585 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07060326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129613 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129608 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07325383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129616 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129542 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07319278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06764910 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129561 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129609 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06764915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06764913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129559 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07141359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07141362 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06899917 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06759605 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129599 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129588 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06899915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129581 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129590 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07319288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129568 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07088279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26129587 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10581153 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581154 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581152 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581156 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581157 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581155 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581158 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422562_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10581151 | - | - |
| Mouse Genome 430 2.0 Array | 1422562_at | - | - |
| Mouse Genome 430 2.0 Array | 1422562_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56437_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56437_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800021548.mm.1 | NM_019662.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021548.mm.1 | NM_019662.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021548.mm.1 | TC0800002807.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422562_at | - | - |
| Mouse Expression Set 430 | 1422562_PM_at | - | - |
| Murine Genome U74v2 Set | 97319_at | - | - |
| Murine Genome U74v2 Set | 97319_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422562_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422562_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422562_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422562_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 884323 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4280129 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4280129 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004280129 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2626242 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2626242 | - | - |
| mouse-6 v1.1 (Illumina) | scl34415.5.1_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4280129 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl34415.5.1_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4280129 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1219106 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1219106 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl34415.5.1_1-S | - | - |

Rrad
Genomic Map