Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900010794.mm.1 |
NM_001243748.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010794.mm.1 |
NM_001243748.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010794.mm.1 |
NM_019713.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010794.mm.1 |
NM_019713.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010794.mm.1 |
TC0900001373.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900010784.mm.1 |
NM_019713.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010784.mm.1 |
NM_019713.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010784.mm.1 |
TC0900001373.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900010792.mm.1 |
NM_001243748.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010792.mm.1 |
NM_001243748.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010792.mm.1 |
NM_019713.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010792.mm.1 |
NM_019713.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010792.mm.1 |
TC0900001373.mm.1 |
MTA TC |