Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200014797.mm.1 |
NM_001252580.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014797.mm.1 |
NM_001252580.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014797.mm.1 |
NM_019729.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014797.mm.1 |
NM_019729.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014797.mm.1 |
TC0200001900.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200014798.mm.1 |
NM_001252580.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014798.mm.1 |
NM_001252580.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014798.mm.1 |
NM_019729.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014798.mm.1 |
NM_019729.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014798.mm.1 |
TC0200001900.mm.1 |
MTA TC |