Assay Details
Gene Symbol: Ranbp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019930.2 | NP_064314.2 | 4 - 5 | 899 | 83 | |
GenBank mRNA | AF006465.1 | - | 5 - 6 | 728 | 83 | |
AK132714.1 | - | 4 - 5 | 378 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
