Assay Details
| Gene Symbol: Slc37a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145960.1 | NP_001139432.1 | 14 - 15 | 1629 | 63 | |
| NM_020258.4 | NP_064654.3 | 14 - 15 | 1629 | 63 | ||
| GenBank mRNA | AF121081.1 | - | 14 - 15 | 1528 | 63 | |
| AK028984.1 | - | 14 - 15 | 1305 | 63 | ||
| AK031045.1 | - | 14 - 15 | 1567 | 63 | ||
| AK079414.1 | - | 14 - 15 | 1305 | 63 | ||
| AK079823.1 | - | 14 - 15 | 1630 | 63 | ||
| AK220151.1 | - | 12 - 13 | 1071 | 63 | ||
| BC038000.1 | - | 14 - 15 | 1300 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc37a2
Genomic Map