Assay Details
| Gene Symbol: Slco1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020495.1 | NP_065241.1 | 1 - 2 | 37 | 70 | |
| GenBank mRNA | AB031959.1 | - | 1 - 2 | 37 | 70 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB031959.1,NM_020495.1 transcripts.
Set Membership: |
|

Slco1b2
Genomic Map