Assay Details
| Gene Symbol: Slco1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020495.1 | NP_065241.1 | 6 - 7 | 670 | 88 | |
| GenBank mRNA | AB031959.1 | - | 6 - 7 | 670 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slco1b2
Genomic Map