Assay Details
| Gene Symbol: U90926 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033483.1 | 4 - 5 | 271 | 82 | ||
| GenBank mRNA | U90926.1 | - | 4 - 5 | 282 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | USO1 vesicle docking factor | Uso1 |
115kDa, TAP, Vdp |
-7.1kb | Anti-sense | Chr. 5 - 92137909 - 92202800 on NCBI Build GRCm38 |
| Downstream |
protein phosphatase, EF hand calcium-binding domain 2 |
Ppef2 | 15.6kb | Sense | Chr. 5 - 92226691 - 92256900 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

U90926
Genomic Map