Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700009091.mm.1 |
TC0700000989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700009091.mm.1 |
NM_021336.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009091.mm.1 |
NM_021336.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009097.mm.1 |
TC0700000989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700009097.mm.1 |
NM_021336.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009097.mm.1 |
NM_021336.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009098.mm.1 |
TC0700000989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700009098.mm.1 |
NM_021336.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009098.mm.1 |
NM_021336.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009095.mm.1 |
TC0700000989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700009095.mm.1 |
NM_021336.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009095.mm.1 |
NM_021336.4 |
Refseq |