Assay Details
| Gene Symbol: Snrpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021336.4 | NP_067311.4 | 3 - 4 | 587 | 102 | |
| GenBank mRNA | AF230356.1 | - | 3 - 4 | 316 | 102 | |
| AK005256.1 | - | 3 - 4 | 379 | 102 | ||
| AK088438.1 | - | 3 - 4 | 362 | 102 | ||
| AK151800.1 | - | 3 - 4 | 380 | 102 | ||
| AK152546.1 | - | 3 - 4 | 380 | 102 | ||
| AK153502.1 | - | 3 - 4 | 380 | 102 | ||
| AK206837.1 | - | 2 - 3 | 150 | 102 | ||
| BC013777.1 | - | 3 - 4 | 354 | 2651704 | 102 | |
| BC029642.1 | - | 3 - 4 | 354 | 3588138 | 102 | |
More Information
Set Membership: |
|

Snrpa1
Genomic Map