Assay Details
| Gene Symbol: Stk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021420.3 | NP_067395.1 | 9 - 10 | 1184 | 74 | |
| GenBank mRNA | AF271360.1 | - | 9 - 10 | 1197 | 74 | |
| AK028838.1 | - | 9 - 10 | 1186 | 74 | ||
| BC054521.1 | - | 9 - 10 | 1210 | 6409342 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stk4
Genomic Map