Assay Details
Gene Symbol: Ngly1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021504.3 | NP_067479.2 | 7 - 8 | 1191 | 132 | |
GenBank mRNA | AF250927.1 | - | 7 - 8 | 1178 | 132 | |
AK028248.1 | - | 7 - 8 | 1192 | 132 | ||
AK148931.1 | - | 7 - 8 | 1194 | 132 | ||
AK169205.1 | - | 7 - 8 | 1179 | 132 | ||
BC028961.1 | - | 7 - 8 | 1145 | 5366023 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
