Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700020118.mm.1 |
NM_001199113.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020118.mm.1 |
NM_001199113.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020118.mm.1 |
NM_001199114.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020118.mm.1 |
NM_001199114.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020118.mm.1 |
NM_001199115.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020118.mm.1 |
NM_001199115.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020118.mm.1 |
NM_001199116.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020118.mm.1 |
NM_001199116.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020118.mm.1 |
NM_022880.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020118.mm.1 |
NM_022880.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020118.mm.1 |
TC1700002152.mm.1 |
MTA TC |